Assay Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249909.2 | XP_008248131.1 | 4395 | 67 | ||
| XM_017338344.1 | XP_017193833.1 | 4244 | 67 | |||
| XM_017338345.1 | XP_017193834.1 | 3559 | 67 | |||
| XM_017338346.1 | XP_017193835.1 | 1368 | 67 | |||
More Information
Set Membership: |
|

ZC3H12D
Genomic Map