Assay Details
| Gene Symbol: DTX3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347057.1 | XP_017202546.1 | 2 - 3 | 421 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DTX3L| Gene Symbol: DTX3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347057.1 | XP_017202546.1 | 2 - 3 | 421 | 59 | |
Set Membership: |
|