Assay Details
| Gene Symbol: DEDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722303.3 | XP_002722349.3 | 644 | 69 | ||
| XM_008249678.2 | XP_008247900.2 | 651 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEDD2| Gene Symbol: DEDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722303.3 | XP_002722349.3 | 644 | 69 | ||
| XM_008249678.2 | XP_008247900.2 | 651 | 69 | |||
Set Membership: |
|