Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270818.2 | XP_008269040.1 | 2 - 3 | 538 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A11| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270818.2 | XP_008269040.1 | 2 - 3 | 538 | 106 | |
Set Membership: |
|