Assay Details
| Gene Symbol: GUCY1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267287.2 | XP_008265509.1 | 6 - 7 | 811 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY1B1| Gene Symbol: GUCY1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267287.2 | XP_008265509.1 | 6 - 7 | 811 | 75 | |
Set Membership: |
|