Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721130.3 | XP_002721176.2 | 344 | 68 | ||
| XM_008274556.2 | XP_008272778.1 | 352 | 68 | |||
| XM_017350634.1 | XP_017206123.1 | 397 | 68 | |||
More Information
Set Membership: |
|

COLEC11
Genomic Map