Assay Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709418.3 | XP_002709464.1 | 3 - 4 | 418 | 75 | |
| XM_017337575.1 | XP_017193064.1 | 3 - 4 | 561 | 75 | ||
More Information
Set Membership: |
|

COMMD8
Genomic Map