Assay Details
| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717289.3 | XP_002717335.1 | 6 - 7 | 1142 | 68 | |
| XM_008268022.2 | XP_008266244.1 | 6 - 7 | 1152 | 68 | ||
| XM_008268023.2 | XP_008266245.1 | 6 - 7 | 1231 | 68 | ||
| XM_017347648.1 | XP_017203137.1 | 5 - 6 | 1606 | 68 | ||
More Information
Set Membership: |
|

MAP3K8
Genomic Map