Assay Details
| Gene Symbol: LOC100343925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718440.3 | XP_002718486.1 | 2 - 3 | 711 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100343925| Gene Symbol: LOC100343925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718440.3 | XP_002718486.1 | 2 - 3 | 711 | 109 | |
Set Membership: |
|