Assay Details
| Gene Symbol: EEF1AKMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259927.2 | XP_008258149.1 | 5 - 6 | 768 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EEF1AKMT1| Gene Symbol: EEF1AKMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259927.2 | XP_008258149.1 | 5 - 6 | 768 | 89 | |
Set Membership: |
|