Assay Details
| Gene Symbol: BLOC1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256607.2 | XP_008254829.1 | 3 - 4 | 357 | 73 | |
| XM_017341922.1 | XP_017197411.1 | 4 - 5 | 508 | 73 | ||
More Information
Set Membership: |
|

BLOC1S1
Genomic Map