Assay Details
| Gene Symbol: KIF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346495.1 | XP_017201984.1 | 7 - 8 | 1237 | 73 | |
| XM_017346496.1 | XP_017201985.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346497.1 | XP_017201986.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346498.1 | XP_017201987.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346499.1 | XP_017201988.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346500.1 | XP_017201989.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346501.1 | XP_017201990.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346502.1 | XP_017201991.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346503.1 | XP_017201992.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346504.1 | XP_017201993.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346505.1 | XP_017201994.1 | 7 - 8 | 1237 | 73 | ||
| XM_017346506.1 | XP_017201995.1 | 7 - 8 | 1237 | 73 | ||
More Information
Set Membership: |
|

KIF17
Genomic Map