Assay Details
| Gene Symbol: MKRN2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260423.2 | XP_008258645.1 | 1 - 2 | 775 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MKRN2OS| Gene Symbol: MKRN2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260423.2 | XP_008258645.1 | 1 - 2 | 775 | 83 | |
Set Membership: |
|