Assay Details
| Gene Symbol: FGF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716514.3 | XP_002716560.1 | 2 - 3 | 1427 | 65 | |
| XM_008266665.2 | XP_008264887.1 | 2 - 3 | 287 | 65 | ||
| XM_008266666.1 | XP_008264888.1 | 3 - 4 | 392 | 65 | ||
More Information
Set Membership: |
|

FGF12
Genomic Map