Assay Details
| Gene Symbol: CHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256976.2 | XP_008255198.1 | 2 - 3 | 449 | 68 | |
| XM_017342057.1 | XP_017197546.1 | 2 - 3 | 449 | 68 | ||
More Information
Set Membership: |
|

CHPT1
Genomic Map