Assay Details
| Gene Symbol: LOC100358177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715799.3 | XP_002715845.1 | 1 - 2 | 113 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002715799.3 transcripts.
Set Membership: |
|

LOC100358177
Genomic Map