Assay Details
| Gene Symbol: KCNA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252358.2 | XP_008250580.1 | 2517 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNA7| Gene Symbol: KCNA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252358.2 | XP_008250580.1 | 2517 | 77 | ||
Set Membership: |
|