Assay Details
| Gene Symbol: GCM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714506.3 | XP_002714552.1 | 6 | 902 | 96 | |
| XM_017345203.1 | XP_017200692.1 | 5 | 828 | 96 | ||
More Information
Set Membership: |
|

GCM1
Genomic Map