Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249625.2 | XP_008247847.1 | 2097 | 81 | ||
| XM_008249626.2 | XP_008247848.1 | 2093 | 81 | |||
| XM_017338208.1 | XP_017193697.1 | 2092 | 81 | |||
| XM_017338209.1 | XP_017193698.1 | 1906 | 81 | |||
More Information
Set Membership: |
|

ZNF500
Genomic Map