Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262333.2 | XP_008260555.1 | 2 - 3 | 261 | 70 | |
| XM_017344707.1 | XP_017200196.1 | 2 - 3 | 190 | 70 | ||
More Information
Set Membership: |
|

CCL28
Genomic Map