Assay Details
| Gene Symbol: LOC100342699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342602.1 | XP_017198091.1 | 1 - 2 | 1892 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017342602.1 transcripts.
Set Membership: |
|

LOC100342699
Genomic Map