Assay Details
| Gene Symbol: LOC100342699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342602.1 | XP_017198091.1 | 6 - 7 | 2925 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100342699| Gene Symbol: LOC100342699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342602.1 | XP_017198091.1 | 6 - 7 | 2925 | 65 | |
Set Membership: |
|