Assay Details
| Gene Symbol: LOC100356015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709593.2 | XP_002709639.1 | 3 - 4 | 501 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100356015| Gene Symbol: LOC100356015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709593.2 | XP_002709639.1 | 3 - 4 | 501 | 99 | |
Set Membership: |
|