Assay Details
| Gene Symbol: LOC4325486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757621.2 | XP_015613107.1 | 3 - 4 | 380 | 133 | |
| XM_015757642.2 | XP_015613128.1 | 3 - 4 | 382 | 133 | ||
| GenBank mRNA | AK065031.1 | - | 3 - 4 | 377 | 133 | |
More Information
Set Membership: |
|

LOC4325486
Genomic Map