Assay Details
| Gene Symbol: LOC4325039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758376.2 | 1 - 2 | 267 | 86 | ||
| GenBank mRNA | AK120577.1 | - | 1 - 2 | 149 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015758376.2 transcripts.
Set Membership: |
|

LOC4325039
Genomic Map