Assay Details
| Gene Symbol: LOC4325039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758376.2 | 4 - 5 | 522 | 104 | ||
| GenBank mRNA | AK120577.1 | - | 4 - 5 | 404 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325039| Gene Symbol: LOC4325039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758376.2 | 4 - 5 | 522 | 104 | ||
| GenBank mRNA | AK120577.1 | - | 4 - 5 | 404 | 104 | |
Set Membership: |
|