Assay Details
| Gene Symbol: LOC4325039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758376.2 | 5 - 6 | 691 | 84 | ||
| GenBank mRNA | AK120577.1 | - | 5 - 6 | 573 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325039| Gene Symbol: LOC4325039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758376.2 | 5 - 6 | 691 | 84 | ||
| GenBank mRNA | AK120577.1 | - | 5 - 6 | 573 | 84 | |
Set Membership: |
|