Assay Details
| Gene Symbol: LOC4329009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771422.2 | XP_015626908.1 | 10 - 11 | 1474 | 89 | |
| GenBank mRNA | AK110791.1 | - | 10 - 11 | 1177 | 89 | |
More Information
Set Membership: |
|

LOC4329009
Genomic Map