Assay Details
| Gene Symbol: LOC4329009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771422.2 | XP_015626908.1 | 8 - 9 | 1217 | 101 | |
| GenBank mRNA | AK110791.1 | - | 8 - 9 | 920 | 101 | |
More Information
Set Membership: |
|

LOC4329009
Genomic Map