Assay Details
| Gene Symbol: LOC4329068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767431.2 | XP_015622917.1 | 2 | 2244 | 117 | |
| XM_015767432.2 | XP_015622918.1 | 2 | 2012 | 117 | ||
| GenBank mRNA | AK058474.1 | - | 1 | 1434 | 117 | |
| AK103308.1 | - | 1 | 2251 | 117 | ||
More Information
Set Membership: |
|

LOC4329068
Genomic Map