Assay Details
| Gene Symbol: LOC4331675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777845.2 | XP_015633331.1 | 10 - 11 | 1139 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4331675| Gene Symbol: LOC4331675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777845.2 | XP_015633331.1 | 10 - 11 | 1139 | 68 | |
Set Membership: |
|