Assay Details
| Gene Symbol: LOC4331675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777845.2 | XP_015633331.1 | 5 - 6 | 819 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4331675| Gene Symbol: LOC4331675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777845.2 | XP_015633331.1 | 5 - 6 | 819 | 140 | |
Set Membership: |
|