Assay Details
| Gene Symbol: LOC4331675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777845.2 | XP_015633331.1 | 8 - 9 | 1001 | 110 | |
| GenBank mRNA | AK073432.1 | - | 8 - 9 | 978 | 110 | |
More Information
Set Membership: |
|

LOC4331675
Genomic Map