Assay Details
| Gene Symbol: LOC4332791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776569.2 | XP_015632055.1 | 11 - 12 | 1394 | 150 | |
| GenBank mRNA | AB042935.1 | - | 11 - 12 | 1399 | 150 | |
| AK066679.1 | - | 11 - 12 | 1401 | 150 | ||
More Information
Set Membership: |
|

LOC4332791
Genomic Map