Assay Details
| Gene Symbol: LOC4332791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776569.2 | XP_015632055.1 | 15 - 16 | 1682 | 92 | |
| GenBank mRNA | AB042935.1 | - | 15 - 16 | 1687 | 92 | |
| AK066679.1 | - | 15 - 16 | 1689 | 92 | ||
More Information
Set Membership: |
|

LOC4332791
Genomic Map