Assay Details
| Gene Symbol: LOC4332791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776569.2 | XP_015632055.1 | 1 - 2 | 241 | 127 | |
| GenBank mRNA | AB042935.1 | - | 1 - 2 | 246 | 127 | |
| AK066679.1 | - | 1 - 2 | 248 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776569.2 transcripts.
Set Membership: |
|

LOC4332791
Genomic Map