Assay Details
| Gene Symbol: LOC4333736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773691.2 | XP_015629177.1 | 1 - 2 | 458 | 85 | |
| GenBank mRNA | AK060516.1 | - | 1 - 2 | 464 | 85 | |
| AK099266.1 | - | 1 - 2 | 465 | 85 | ||
More Information
Set Membership: |
|

LOC4333736
Genomic Map