Assay Details
| Gene Symbol: LOC4333736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773691.2 | XP_015629177.1 | 3 - 4 | 960 | 59 | |
| GenBank mRNA | AK060516.1 | - | 3 - 4 | 966 | 59 | |
| AK099266.1 | - | 3 - 4 | 967 | 59 | ||
More Information
Set Membership: |
|

LOC4333736
Genomic Map