Assay Details
| Gene Symbol: LOC9267465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772002.2 | XP_015627488.1 | 1 - 2 | 918 | 130 | |
| GenBank mRNA | AK071766.1 | - | 1 - 2 | 604 | 130 | |
More Information
Set Membership: |
|

LOC9267465
Genomic Map