Assay Details
| Gene Symbol: LOC4331005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771853.2 | XP_015627339.1 | 1 - 2 | 288 | 129 | |
| XM_015771854.2 | XP_015627340.1 | 1 - 2 | 288 | 129 | ||
| XM_015771856.2 | XP_015627342.1 | 1 - 2 | 288 | 129 | ||
| GenBank mRNA | AK105305.1 | - | 1 - 2 | 279 | 129 | |
More Information
Set Membership: |
|

LOC4331005
Genomic Map