Assay Details
| Gene Symbol: LOC4331005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771853.2 | XP_015627339.1 | 3 - 4 | 780 | 82 | |
| XM_015771854.2 | XP_015627340.1 | 3 - 4 | 780 | 82 | ||
| XM_015771856.2 | XP_015627342.1 | 3 - 4 | 780 | 82 | ||
| GenBank mRNA | AK105305.1 | - | 3 - 4 | 771 | 82 | |
More Information
Set Membership: |
|

LOC4331005
Genomic Map