Assay Details
Gene Symbol: LOC4331986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015773227.2 | XP_015628713.1 | 3 - 4 | 2057 | 96 | |
GenBank mRNA | AK121042.1 | - | 3 - 4 | 2024 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
