Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 32 - 33 | 4481 | 85 | |
| GenBank mRNA | AK099757.1 | - | 11 - 12 | 1376 | 85 | |
More Information
Set Membership: |
|

LOC4332045
Genomic Map