Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 34 - 35 | 4630 | 77 | |
| GenBank mRNA | AK099757.1 | - | 13 - 14 | 1525 | 77 | |
| AK120639.1 | - | 10 - 11 | 1055 | 77 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map