Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 43 - 44 | 5726 | 77 | |
| GenBank mRNA | AK099757.1 | - | 22 - 23 | 2621 | 77 | |
| AK120639.1 | - | 19 - 20 | 2151 | 77 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map