Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 48 - 49 | 6448 | 71 | |
| GenBank mRNA | AK099757.1 | - | 27 - 28 | 3343 | 71 | |
| AK120639.1 | - | 24 - 25 | 2873 | 71 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map