Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 30 - 31 | 4260 | 149 | |
| GenBank mRNA | AK099757.1 | - | 9 - 10 | 1155 | 149 | |
| AK120639.1 | - | 6 - 7 | 674 | 149 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map