Assay Details
| Gene Symbol: LOC4334126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776502.2 | XP_015631988.1 | 7 - 8 | 959 | 77 | |
| GenBank mRNA | AK061386.1 | - | 7 - 8 | 857 | 77 | |
| AK066914.1 | - | 4 - 5 | 248 | 77 | ||
| AK072597.1 | - | 7 - 8 | 934 | 77 | ||
More Information
Set Membership: |
|

LOC4334126
Genomic Map