Assay Details
| Gene Symbol: LOC4335795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061508.1 | - | 7 - 8 | 1388 | 118 | |
| AK101471.1 | - | 7 - 8 | 2057 | 118 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4335795| Gene Symbol: LOC4335795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061508.1 | - | 7 - 8 | 1388 | 118 | |
| AK101471.1 | - | 7 - 8 | 2057 | 118 | ||
Set Membership: |
|