Assay Details
| Gene Symbol: LOC4336300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780153.2 | XP_015635639.1 | 3 - 4 | 530 | 122 | |
| GenBank mRNA | AK065553.1 | - | 4 - 5 | 362 | 122 | |
| AY224450.1 | - | 3 - 4 | 318 | 119 | ||
More Information
Set Membership: |
|

LOC4336300
Genomic Map